Learn More
Thermo Scientific™ Nuts for Capillary GC Columns Connections
Catalog No. 03050223
Change view
Click to view available options
Material:
Brass
Catalog No. Material
03050223 Brass
1 options
Catalog No. 03050223 Supplier Thermo Scientific™ Supplier No. 290BT239
Use Thermo Scientific™ nuts with compatible ferrules to eliminate leaks and connect your capillary GC columns to your instrument.
Use Thermo Scientific™ nuts with compatible ferrules to eliminate leaks and connect your capillary GC columns to your instrument. Thermo Scientific™ ferrules are available in three different materials - 100% Graphite, 15% Graphite / 85% Vespel, or SilTite Metal - and various dimensions to match your instrument and capillary column inner diameter. Use the nut that is compatible with your ferrule.
Specifications
Type Capillary Column Nut
For Use With (Equipment) Graphite/Vespel Ferrules
Material Brass
Description Brass Nut For 15% Graphite/85% Vespel Ferrules