Learn More
Thermo Scientific™ Nutrient Broth (Dehydrated)
Cultivate fastidious pathogens and other microorganisms with this nutritious medium.
$324.60 - $324.60
Specifications
Form Powder
Product Type Dehydrated Culture Media
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
OXCM0001B
View Documents
Thermo Scientific™
CM0001B
500 g
Each for $324.60
Only null left
Description
Description
Cultivate fastidious pathogens and other microorganisms with nutritious Thermo Scientific™ Oxoid™ Nutrient Broth (Dehydrated). This general purpose medium contains beef extracts combined with peptone, glucose, yeast extracts and sodium chloride to provide the nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.
A general purpose fluid medium for the cultivation of microorganisms not exacting in their nutritional requirements.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
Powder
Dehydrated Culture Media
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
For Laboratory Use Only