missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ MOPS SDS Running Buffer (Powder)

Catalog No. NP000205
Change view
Click to view available options
Quantity:
5 Packets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP000205 5 Packets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP000205 Supplier Invitrogen™ Supplier No. NP000205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen NuPAGE MOPS SDS Running Buffer (Powder) is formulated for use with Bis-Tris gels to resolve medium- to large-sized proteins by providing slower SDS-PAGE separations. Supplied as a powder for flexible preparation, room temperature storage, and a long 2-year shelf life.

Invitrogen NuPAGE MOPS SDS Running Buffer (Powder) is formulated for use with Bis-Tris gels to improve the resolution and band sharpness of medium- to large-sized proteins. Compared to MES-based buffers, the MOPS buffer system supports reduced protein migration rates for enhanced separation performance. Supplied as a dry powder, this running buffer enables flexible preparation, room temperature storage, a two-year shelf life, and is designed to be a convenient and reliable choice for routine electrophoresis workflows.

Features and benefits include:

  • Improved resolution of medium- to large-sized proteins—slower protein migration with MOPS buffer enhances separation and band sharpness compared to faster MES-based buffers
  • Optimized for Bis-Tris gel workflows—designed for use with Bis-Tris gels including both NuPAGE Bis-Tris and Bolt Bis-Tris Plus gels
  • Powder format for flexible preparation—prepare fresh running buffer as needed to support consistent electrophoresis performance and simplify laboratory workflows
  • Room temperature storage and extended shelf life—powder format enables convenient storage with a two-year shelf life for inventory efficiency
  • Reliable performance for routine SDS-PAGE—supports consistent and reproducible results across common protein electrophoresis applications when used under recommended conditions

Specifications

Color White
Format Powder
Gel Type Bis-Tris
Formulation MOPS SDS Running Buffer
For Use With (Application) SDS-PAGE
Quantity 5 Packets
Storage Requirements Room Temperature
Sufficient For Preparing 5 L Running Buffer
Product Line NuPAGE
Product Type SDS Running Buffer
For Use With (Equipment) Electrophoresis Chambers
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.