Learn More
Thermo Scientific™ Nunc™ Storage Vials with Caps
Assure safe centrifugation and storage of PCR reagents, restriction enzymes, and other diagnostic and biochemical reagents or samples by using these storage vials with caps.
Supplier: Thermo Scientific™ 264300
Description
Assure safe centrifugation and storage of PCR reagents, restriction enzymes and other biochemical reagents and samples by using Thermo Scientific™ Nunc™ Storage Vials with Caps. Available in 0.5mL and 2.0mL sizes with frosted writing area, the vials have excellent chemical resistance with leakproof closure.
Features:
- Made from high-purity polypropylene copolymer with excellent chemical resistance
- Leakproof high-density polyethylene screw closure
- Centrifuged to a maximum of 13,000 x g
- Temperature range: -40°C to 95°C One full turn removes the ergonomic closure
- RNase/DNase-free
Ordering Information:
Racks 264263 and 270972 are compatible with these vials. Color coders available: 254416, 264335, 264408, 264440, 264459, 264467, 264491.
Specifications
| PPCO | |
| 2.2 mL | |
| 500 | |
| Nunc | |
| Polyethylene | |
| 500 Cs. | |
| Storage Vial with Cap |
| Sterile | |
| 500 | |
| 2.0 mL | |
| 2.2 mL | |
| Screw Cap | |
| Certificate of Analysis (COA), Certificate of Conformance (COC) |