missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
GREENER_CHOICE

Catalog No. p-4525142
Change view
Click to view available options
Material:
Aluminum
Non-woven Rayon
Plastic
Polyester
Polyethylene
Polyolefin
Polypropylene
PVC-coated Polypropylene
Vinyl
For Use With (Application):
Breathable, Cell culture
Cell culture, sterile applications
Cell growth, Bacterial growth
Consistent seal
Crystallography and genotyping
General purpose, ELISA
General purpose, sample detection
Low autofluorescence, ELISA
Luminescence, photosensitive samples
PCR
PCR, fluorescence assays, microscopy
PCR, long-term storage
PCR, qPCR, ELISA, Protein crystallography
PCR, storage in 384 well plates
PCR, storage in 96-well plates
Short-term storage, ELISA
Short-term storage, ELISA, minimizes evaporation and cross contamination
Ventilating and storing cell cultures
Certified RNase/DNase-free:
No
Yes
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material For Use With (Application) Certified RNase/DNase-free
12565398 Aluminum PCR, storage in 96-well plates Yes
12565399 Aluminum PCR, storage in 384 well plates Yes
12565492 Polypropylene PCR Yes
12565491 Polypropylene PCR, fluorescence assays, microscopy Yes
12565512 Polyethylene Low autofluorescence, ELISA No
12565513 Polyolefin PCR, qPCR, ELISA, Protein crystallography Yes
12565171 Vinyl Luminescence, photosensitive samples No
1256571 Polyester Cell culture, sterile applications No
1256790 Polyester General purpose, sample detection No
12566914 Polyester General purpose, ELISA Yes
1256705 Non-woven Rayon Breathable, Cell culture No
12249720 Non-woven Rayon Cell growth, Bacterial growth Yes
12565463 Plastic Consistent seal No
12565475 Aluminum PCR, long-term storage Yes
1424518 Polyester Short-term storage, ELISA No
14245192B Polyester Short-term storage, ELISA, minimizes evaporation and cross contamination No
AB0718 Polypropylene Ventilating and storing cell cultures No
AB0580 PVC-coated Polypropylene Crystallography and genotyping No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 12565398 Supplier Thermo Scientific™ Supplier No. 232698
Only null left

Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.

Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.

  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)

Specifications

Color Foil
Solvent Or Alcohol Resistant Life Science Reagent Resistant
Description AL SEAL TAPE FOR 96 WELL PLT
Material Aluminum
Sterility Non-sterile
Autoclavable Not Autoclavable
No. of Wells 96
No. per Case 100
No. per Pack 100
Evaporation Barrier Malleable allows for visual verification of seal security, minimizes evaporation and cross contamination
For Use With (Application) PCR, storage in 96-well plates
Content And Storage General; Long term
Product Type Adhesive Seal
Product Line Nunc
Adhesive Acrylic
Certified RNase/DNase-free Yes
Pierceable Yes
Temperature Range (Metric) -80°C to +120°C
Well Shape All well shapes
Plate Compatibility All Microplates
Incubation Conditions General
Peelable No
Show More Show Less