missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
Adhesive Plate Seals provide an effective seal. It features an easy-removal non-adhesive tab and can withstand temperatures between -40°C to 80°C.
Highlights
- Non-adhesive tab for easy removal of seal
- Temperature range: -40°C to 80°C
Specifications
Specifications
| Color | Clear |
| Solvent Or Alcohol Resistant | Life Science Reagent Resistant |
| Description | Gas Permeable Adhesive Seal |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| No. of Wells | 96 |
| No. per Case | 50 |
| No. per Pack | 50 |
| For Use With (Application) | Ventilating and storing cell cultures |
| Show More |