missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils

Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
Protect microplate well contents with the Thermo Scientific™ Mylar™ Plate Sealer for Microtiter™ Plates, which assures a tight seal. Designed for use when sealing microtiter plates.
Specifications
Specifications
| Color | Clear |
| Solvent Or Alcohol Resistant | Life Science Reagent Resistant |
| Description | SEALER, MYLAR, 100/CS |
| Material | Polyester |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| No. of Wells | 96 |
| No. per Case | 100 |
| No. per Pack | 100 |
| Dimensions (L x W) | 83 x 124 mm |
| Show More |