missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Press Out Tool for Cryobank and Bank-It™ Vial Systems

Catalog No. 12565003
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12565003 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12565003 Supplier Thermo Scientific™ Supplier No. 374009

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Minimize freeze/thaw by using the Thermo Scienctific™ Nunc™ Press Out Tool for Cryobank and Bank-It™ Systems, which allows for rapid retrieval of Cryobank vials.

Locate and remove a desired tube quickly and easily, without allowing frozen samples to thaw by using the Thermo Scientific™ Nunc™ Press Out Tool for Cryobank and Bank-It™ Vial Systems.

Features:

  • Selected vials are released by levers and can be easily harvested
  • Selected position can be chosen based on data base output
  • Pegs can be placed and checked while the sample/neighboring samples remain cold
  • Cut-off corner ensures the rack is orientated correctly

Specifications

Description Nunc Cryobank and Bank-it Rack, Tube Press out Tool, 1 Each
Vial Type Cryobank
Color Silver
For Use With (Equipment) Cryobank and Bank-It Systems
Quantity 1 Ea.
Type Tube Press Out Tool