Learn More
Thermo Scientific™ Nunc™ Non-Treated Multidishes
Perform suspension cell cultures with Thermo Scientific™ Nunc™ Non-Treated Multidishes.
Supplier:  Thermo Scientific™ 179820
 View more versions of this product
Catalog No. 12566301
Only null left
Description
Description
Perform suspension cell cultures with Thermo Scientific™ Nunc™ Non-Treated Multidishes.
  • Flat-bottom round wells, with lid
  • Excellent optical properties
  • Raised well rims to lower the risk of cross-contamination
  • Lid with notched corners for good fit and easy orientation
  • Non-pyrogenic
Specifications
Chemical Identifiers
Multi-Well Plate
Non-treated
Lid
Sterile
Flat Bottom (F-well)
Specifications
Multi-Well Plate
4
Sterile
Flat Bottom (F-well)
120
66 mm
1 mL (Well)
1 mL
Non-treated
Lid
4
66 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.