missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Multidishes with Nunclon™ Supra Surface

Catalog No. 12565674
Change view
Click to view available options
No. of Wells:
12
24
48
6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
12565674 12
12565675 6
12565673 24
12565672 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12565674 Supplier Thermo Scientific™ Supplier No. 140681
Only null left

Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.

Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc Multidishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

Specifications

Lid With Lid
Product Line Nunc
Color Clear
Disposable Single-use
Material Polystyrene
No. of Wells 12
Sterility Sterile
Surface Treatment Nunclon Supra
Well Shape Flat Bottom (F-well)
No. per Case 75 Dishes
No. per Pack 1 Dish
Show More Show Less

For Laboratory Use