missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 96 Black Plate, Non-Treated Coverglass Surface, No Lid, Non-Sterile, Pack of 5
Color:Black, Clear Bottom; Sterility; Non-sterile; Lid, Without Lid; Type: Optical Coverglass Base
Supplier: Thermo Scientific™ 265300
Description
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with the Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
Features:
- Background, autoflorescence and crosstalk are improved
- Black microplates for florescence measurements, with minimum back-scattered light and background fluorescence
- White microplates for luminescence measurements, with maximum reflection and minimal autoluminescence
- Cell culture surface is ideal for cell-based assays
- CC2 surface modification closely mimics Poly-D-Lysine and is superior for attachment and growth of fastidious cells
- Customized bar coding available
Specifications
| Custom Barcoding Available | |
| Without Lid | |
| 5/Pk., 30/Cs. | |
| Black | |
| Polystyrene | |
| Non-sterile | |
| Flat Bottom (F-well) | |
| 5 |
| All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
| 8 x 12 | |
| Nunc | |
| Optical Coverglass Base | |
| 96 | |
| Non-treated | |
| 30 | |
| Case of 30 |