missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Color: White; Sterility: Sterile; Lid: With Lid
$1,700.29 - $4,113.84
Specifications
| Barcode | Custom Barcoding Available |
|---|---|
| Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
| Certified | Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility |
| Lid | With Lid |
| Array | 8 x 12 |
| Catalog Number | Mfr. No. | No. of Wells | No. per Pack | No. per Case | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Wells | No. per Pack | No. per Case | Price | Quantity & Availability | |||
|
1256600
|
Thermo Scientific™
N136101 |
96 | 1 Microplate | 50 Microplates |
Case of 50 Each for $1,700.29
|
|
|||
|
1256626
|
Thermo Scientific™
136102 |
96 | 10 Microplates | 160 Microplates |
Case of 160 Each for $4,113.84
|
|
|||
Description
Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ White Polystyrene Plate. White plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics.
- Background, autoflorescence and crosstalk are improved when you use Nunc White microplates.
- White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
- Working volume range: 50 to 250μL/well
- Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
- Customized bar coding is available
Specifications
| Custom Barcoding Available | |
| Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility | |
| 8 x 12 | |
| White | |
| Low Flange Pinchbar | |
| 400 μL | |
| 50 to 200 μL | |
| Nunclon Delta |
| All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
| With Lid | |
| Nunc | |
| Microplate | |
| Polystyrene | |
| 96 | |
| Sterile | |
| Flat Bottom (F-well) |