missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Black, sterile microplates with lid. Ideal for fluorescence measurements, with minimum back-scattered light and background fluorescence.
Supplier: Thermo Scientific™ 137103
Description
Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ Black and White Polystyrene Plate. Black and white plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics. Available with or without cell culture treatment.
- Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
- Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
- White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
- Working volume range: 50 to 250μL/well
- Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
- Customized bar coding is available
Chemical Identifiers
| Custom Barcoding Available | |
| Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility | |
| 8 x 12 | |
| Black | |
| Polystyrene | |
| 96 | |
| Sterile | |
| Flat Bottom (F-well) |
| All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
| With Lid | |
| Nunc | |
| Microplate | |
| 400 μL | |
| 50 to 250 μL/well | |
| Nunclon Delta |
Specifications
| Custom Barcoding Available | |
| Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility | |
| 8 x 12 | |
| Nunc | |
| Microplate | |
| 400 μL | |
| 50 to 250 μL/well | |
| Nunclon Delta | |
| 160 | |
| 160/Cs. |
| All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
| With Lid | |
| 10 Pk., 160 Cases | |
| Black | |
| Polystyrene | |
| 96 | |
| Sterile | |
| Flat Bottom (F-well) | |
| 10 |