missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: Without Lid
Supplier: Thermo Scientific™ 156545
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Specifications
| Custom Barcoding Available | |
| Without Lid | |
| 8 x 12 | |
| Nunc | |
| Microplate | |
| Polystyrene | |
| 96 | |
| Nunclon Delta | |
| 180 Microplates | |
| 180/Cs. |
| All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
| 96F well plate, Nunclon Delta, clear, without lid, Sterile (Pack of 10, Case of 180) | |
| 10/Pk., 180/Cs. | |
| Clear | |
| High Flange | |
| 400 μL | |
| Sterile | |
| Flat Bottom (F-well) | |
| 10 Microplates |