missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 96 Well Plate, Non-Treated, Surface, No Lid, Non-sterile, Pack of 10
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Specifications
Specifications
| Barcode | Custom Barcoding Available |
| Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
| Lid | Without Lid |
| Description | 96F well plate, Non-Treated, clear, without lid, Non-sterile (Pack of 10, Case of 180) |
| Array | 8 x 12 |
| Packaging | 10 Pk., 180 Case |
| Product Line | Nunc |
| Color | Clear |
| Product Type | Microplate |
| Material | Polystyrene |
| Show More |