Learn More
Thermo Scientific™ Nunc™ 96-Well Optical-Bottom Microplate, black, TC surface
Catalog No. 1256670
Change view
Click to view available options
Quantity:
30/Cs.
Catalog No. Quantity
1256670 30/Cs.
1 options
Catalog No. 1256670 Supplier Thermo Scientific™ Supplier No. 165305
Only null left
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with Nunc MicroWell 384-well optical-bottom plates, featuring a polystyrene black upper structure with a polystyrene film at the bottom of the plate.
Thermo Scientific Nunc MicroWell 96-well optical-bottom plates with polymer base have a solid polystyrene black upper structure with a polystyrene film at the bottom of the plate. The plates are optimized for fluorescence and luminescence in cell culture and microscopic applications.
Black polystyrene upper structure with polymer bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications.
  • Background, autoflorescence, and crosstalk are improved when you use Nunc black microplates
  • Black microplates are recommended for florescence measurements with minimum back-scattered light and background fluorescence
  • Also available in white; recommended for luminescence measurements with maximum reflection and minimal autoluminescence
  • Polymer base plate
  • Working range: 50–200 μL/well
  • Customized bar coding is available
Color: black with clear bottom
Sterility: sterile
Lid: with lid
Type: optical polymer base
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Lid With Lid
Array 8 x 12
Packaging 10 Pk., 30 Cases
Product Line Nunc
Color Black, Clear Bottom
Product Type Optical Polymer Base
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 50 to 200 mL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
No. per Case 30
No. per Pack 10
Quantity 30/Cs.
Show More Show Less