missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Microscope Slides

Catalog No. 1256514
Change view
Click to view available options
Material:
Permanox
Polystyrene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
1256514 Polystyrene
1256513 Permanox
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1256514 Supplier Thermo Scientific™ Supplier No. 960004
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Exchange glass slides with Thermo Scientific™ Nunc™ Microscope Slides, which offer greater cell attachment.

Achieve similar cell attachment and growth as standard cell culture flasks and dishes with the Thermo Scientific™ Nunc™ Plastic Microscope Slides. Available in either polystyrene or Permanox plastics.

  • Surface treatment of the plastic slides ensures optimal cell attachment and growth, similar to that in the regular cell culture vessels
Warranty: 90 days

Specifications

Length (Metric) 75 mm
Type Microscope Slide
Well Count 0
Frosted 1 End
Precleaned Yes
No. per Case 100/Cs.
No. per Pack 20 Pk.
Material Polystyrene
Width (Metric) 25 mm
Sterility Sterile
Color Clear
Coating Cell-Culture Treated
Description Polystyrene microscope slide
Dimensions (L x W) 75 x 25 mm
Show More Show Less
Which fixation reagents can I use with Permanox plastic?

We have a Technical Bulletin (link below) describing fixation reagents that are compatible with Permanox plastic: https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/D19581.pdf

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.