missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Microplate Lids

Catalog No. 125667
Change view
Click to view available options
Plate Compatibility:
384-well Plate
384/1536-well Plate, OmniTray
96-well Plate
All Microplates
Sterility:
Non-sterile
Sterile
No. per Pack:
1
20
25
5
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plate Compatibility Sterility No. per Pack
125667 384/1536-well Plate, OmniTray Non-sterile 20
12565476 All Microplates Sterile 20
12565478 All Microplates Non-sterile 20
12565462 All Microplates Non-sterile 20
12565748 All Microplates Non-sterile 25
12565749 All Microplates Sterile 25
1256569 96-well Plate Non-sterile 5
1256570 96-well Plate Sterile 1
12565603 384-well Plate Sterile 1
12565604 384-well Plate Non-sterile 20
12565610 384-well Plate Sterile 20
12565477 96-well Plate Sterile 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 125667 Supplier Thermo Scientific™ Supplier No. 448143

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.

Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.

  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market

Specifications

Color White
Dimensions (L x W x H) 5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm)
Material Polystyrene
Sterility Non-sterile
No. of Wells 384/1536
Height (Metric) 7.9 mm
Length (English) 5 in.
Length (Metric) 127 mm
Condensation Rings No
Height (English) 0.3 in.
No. per Case 120
No. per Pack 20
Evaporation Barrier No
Width (English) 3.35 in.
Width (Metric) 85 mm
Format Low Profile
Product Type Plate Cover
Temperature Range (Metric) -20°C to +80°C
Well Shape All well shapes
Plate Compatibility 384/1536-well Plate, OmniTray
Cut-off Corners Long Side
Show More Show Less