missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Microplate Lids
GREENER_CHOICE

Catalog No. 12565603
Change view
Click to view available options
Plate Compatibility:
384-well Plate
384/1536-well Plate, OmniTray
96-well Plate
All Microplates
Sterility:
Non-sterile
Sterile
Condensation Rings:
No
Yes
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plate Compatibility Sterility Condensation Rings
12565603 384-well Plate Sterile No
12565476 All Microplates Sterile No
12565478 All Microplates Non-sterile No
12565462 All Microplates Non-sterile No
12565748 All Microplates Non-sterile Yes
12565749 All Microplates Sterile Yes
1256569 96-well Plate Non-sterile Yes
1256570 96-well Plate Sterile Yes
12565604 384-well Plate Non-sterile No
12565610 384-well Plate Sterile No
12565477 96-well Plate Sterile Yes
125667 384/1536-well Plate, OmniTray Non-sterile No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 12565603 Supplier Thermo Scientific™ Supplier No. 264611
Only null left

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.

Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.

  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market

Specifications

Color Clear
Dimensions (L x W x H) 5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm)
Material Polystyrene
Sterility Sterile
No. of Wells 384
Height (Metric) 7.9 mm
Length (English) 5 in.
Length (Metric) 127 mm
Condensation Rings No
Height (English) 0.3 in.
No. per Case 60
No. per Pack 1
Dimensions (L x W) 5 x 3.35 in. (127 x 85 mm)
Evaporation Barrier Yes
Width (English) 3.35 in.
Width (Metric) 85 mm
Product Type Plate Lid
Product Line Nunc
Adhesive None
Temperature Range (Metric) -20°C to +80°C
Well Shape All well shapes
Plate Compatibility 384-well Plate
Cut-off Corners Long Side
Show More Show Less