missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Latch Rack for 1.8 mL Externally-Threaded Universal Tubes
GREENER_CHOICE

Catalog No. p-9576000
Change view
Click to view available options
Barcode:
2D, linear and human readable
None
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Barcode
12590009 None
12570247 2D, linear and human readable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12590009 Supplier Thermo Scientific™ Supplier No. 331830
Only null left

Versatile tubes for use inside a large storage system, or in smaller scale for traditional cryostorage in dewars.

Use versatile Thermo Scientific™ Nunc™ 1.8 mL Externally-threaded Universal Tubes inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars. Available racked or in bulk, with 2D barcodes or non-coded.

Scalable

  • Tubes and rack are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cryostorage boxes made for 10x10 arrays
  • External threads limit contamination
  • Automation-pickable in Universal Latch Rack

Innovative Universal Cap Design

  • Single-piece construction with molded-in gasket
  • Cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen

Secure Tracking

  • Available with a permanently-bonded, unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
  • 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
  • Scan the tube to access sample information and identify the contents
  • 374502NOV is featured with 1D (linear) barcode and human readable on the side that matches 2D barcode on the bottom of the tube

Stringent Quality Control

  • Every 2D barcoded storage tube is scanned to ensure readability
  • Barcodes are checked against database of previously-assigned codes to prevent duplicates
  • Every tube has a Sterility Assurance Level of 106 and is made with class VI medical grade resins

Specifications

Barcode None
Capacity (Metric) 1.8 mL (Tube)
Description Nunc latch rack
Color Beige
Type Latch Rack
For Use With (Equipment) Cryobank
Holds Externally-Threaded Universal Tubes
Includes 10 empty racks
Quantity 1/Pk., 10/Cs.
Sterility Non-sterile