Learn More
Thermo Scientific™ Nunc™ Lab-Tek™ Flask on Slide
Catalog No. 1256523
Change view
Click to view available options
Material:
Glass
Polystyrene
Description:
Flaskette
SlideFlask
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Description
1256523 Glass Flaskette
12565112 Polystyrene SlideFlask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1256523 Supplier Thermo Scientific™ Supplier No. 177453
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Grow cell cultures directly on slides with the Thermo Scientific™ Nunc™ Lab-Tek™ Flask on Slide, ideal for single-cell autoradiography and immunofluorescence.
Culture cells directly on microscope slides with the Thermo Scientific™ Nunc™ Lab-Tek™ Flask on Slide, ideal for single-cell autoradiography and single-cell immunofluorescence.
SlideFlask Chamber (shown with black cap in photo)
  • Employs tissue culture-treated polystyrene slide attached to 18 × 50mm flask structure via ultrasonic weld—allowing for long-term, leak-free incubation
  • Upper flask structure snaps away easily after culture—freeing slide for microscopic examination
  • Nunclon Delta certified
  • Available Coverglass (171862)
The Flaskette Chamber (shown with white cap)
  • Glass slide with a silicone gasket and a 20 × 50mm upper flask structure
  • Can be sealed to exclude air—useful when embedding cells in resin for electron microscopy
  • For research use only
Specifications
Wells 1
Color Clear
Culture Area 10.0 cm2
Description Flaskette
Surface Treatment Cell Culture
Material Glass
Volume (Metric) Working 2.5 to 5.0 mL
Includes With white solid cap, non-vented
No. per Pack 16
Sterility Sterile
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.