missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Immuno TSP Lids

Catalog No. 12565143
Change view
Click to view available options
Quantity:
Case of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12565143 Case of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12565143 Supplier Thermo Scientific™ Supplier No. 445497
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Simplify hybridoma screening with Thermo Scientific™ Nunc™ Immuno TSP lids for antibody production. Use for bacterial inoculations of 96-well plates.

Simplify hybridoma screening with Thermo Scientific™ Nunc™ Immuno TSP lids for antibody production. Use in conjunction with the Thermo Scientific™ Nunc™ OmniTray™ for bacterial inoculations of 96-well plates. Nunc Immuno TSP Lids feature uniform adsorption properties for efficient screening.

  • Ideal for screening of hybridoma cultures for antibody production
  • For hybridoma screening mounted in the OmniTray
  • Uniform adsorption properties
  • 96-pin lid for bacterial inoculations of 96-well plates

Specifications

Color Clear
Description Inoculation Lid
Material Polystyrene
Sterility Sterile
Length (Metric) 127.4 mm
No. per Case 50
No. per Pack 1
Quantity Case of 50
Width (Metric) 85.3 mm
For Use With (Application) Hybridoma screening
Product Type Plate Lid
Product Line Nunc
Plate Compatibility 96-well Plate, OmniTray
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.