missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ FluoroNunc™/LumiNunc™ 96-Well Plates

Catalog No. p-4525228
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
12565290A White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12565290A Supplier Thermo Scientific™ Supplier No. 437796
Only null left

Use these well plates for quantitative and qualitative solid phase immunoassays (e.g., FLISA) and binding assays.

Use Thermo Scientific™ Nunc FluoroNunc/LumiNunc 96-Well Solid plates for quantitative and qualitative solid phase immunoassays (e.g., FLISA) and binding assays. Thermo Scientific Nunc Immuno plates offer lot-to-lot reliability, well-to-well reproducibility, and are backed by industry-leading quality standards and certifications.

Features:

  • Plate has SBS footprint—fits standard equipment
  • Available with flat-bottom or C-shaped wells
  • Choice of PolySorp™ or MaxiSorp™ surface
  • Offered as black, white or clear plates
  • Certified binding homogeneity/reproducibility

Choice of Surface:

  • PolySorp - Hydrophobic surface preferentially binds biomolecules that have hydrophobic domains.

  • MaxiSorp - Hydrophilic surface is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character. Available with or without lot specific.

  • Performance testing via an IgG binding assay (well-to-well %CV less than 5% for clear plates and less than 10% for colored plates). Tested items come with a certificate.

White:

  • Gives maximum reflection
  • Minimum autofluorescence and autoluminescence

Black:

  • Minimizes background in fluorescence reading
  • Minimizes back-scattered light
  • Minimizes crosstalk

Clear:

  • For time-resolved fluorescence

Compatible with:

Compatible with lids for MicroWell™ plates, sealing tape and breathable membrane

Specifications

Binding Property Hydrophilic
Certifications/Compliance Certified for the absorption of IgG; well to well %CV of less than 10%.
Format Standard
Lid With Lid
Culture Area 2.5 cm2
Detection Method Chemiluminescence
Array 8 x 12
Packaging 60 Cases
Binding Type Adsorption
Product Line Nunc
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 250 μL
Surface Treatment MaxiSorp
Well Shape Round, C Bottom
No. per Case 60
No. per Pack 5
Plate Blocking None
Target Molecule Antibodies (Glycoproteins)
Show More Show Less