missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate

Description
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
Specifications
Specifications
| Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
| Certified | Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility |
| Lid | Without Lid |
| Culture Area | 0.35 cm2/well |
| Array | 8 x 12 |
| Packaging | 1/Pk., 50/Cs. |
| Product Line | Nunc |
| Color | Clear |
| Product Type | Microplate |
| Skirt Style | Low Flange |
| Show More |