Learn More
Thermo Scientific™ 96 Well Edge Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 1
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate.
Supplier: Thermo Scientific™ 267576
Description
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
Specifications
| All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
| 0.35 cm2/well | |
| 1/Pk., 50/Cs. | |
| Clear | |
| Low Flange | |
| 400 μL | |
| Sterile | |
| Flat Bottom (F-well) | |
| 50 |
| Without Lid | |
| 8 x 12 | |
| Nunc | |
| Microplate | |
| Polystyrene | |
| 96 | |
| Non-treated | |
| Less waste, fewer resources used | |
| 1 |