Learn More
Thermo Scientific™ Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface
Description
Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.
Features of Nunc EasYFlask flasks with Nunclon Supra surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years
Specifications
Specifications
| Certifications/Compliance | Non-cytotoxic, Non-pyrogenic, USP Class VI |
| Description | Nunc EasYFlask, 25 cm² |
| Material | Polystyrene |
| For Use With (Application) | General cell culture, cell therapy research, culture of fastidious adherent cells |
| Neck Style | Angled |
| No. per Case | 200 Flasks |
| No. per Pack | 10 Flasks |
| Product Type | Cell Culture Flask |
| Sterility | Sterile |
| Class | Class VI |
| Show More |
For Laboratory Use