Learn More
Thermo Scientific™ Nunc™ EasyFill™ Cell Factory™ System
Save valuable space with Thermo Scientific™ Nunc™ EasyFill™ Cell Factory system. Each 10-layer, single-use system holds the equivalent of 36 T175 flasks.
Supplier:  Thermo Scientific™ 140360
 View more versions of this product
Catalog No. 12-567-302
Only null left
Explore available promotions
Description
Description
Save valuable space with Thermo Scientific™ Nunc™ EasyFill™ Cell Factory system. Each 10-layer, single-use system holds the equivalent of 36 T175 flasks. The linear multi-layer cell culture format is easy to use and promotes easy scalability, ideal for both research and commercial-scale applications.
Features:
  • 'Out of the box' system
  • Large, vented screw closure
  • Versatile port design for pouring and aseptic filling
Benefits:
  • Fill and empty five times faster than T175 flasks
  • No accessories required with EasyFill Cell Factory systems
To Get Started:
Simply pour the media directly into the EasyFill Cell Factory system's large opening.
Warranty: 90 days
Specifications
Specifications
EasyFill Cell Factory
1 Ea.
Less waste, renewable material
Vented
4
4/Cs.
Cell Factory System
Sterile
2528 cm²
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories
Promotions
Promotions
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.