Learn More
Thermo Scientific™ Nunc™ EasyFill™ Cell Factory™ System
Save valuable space with Thermo Scientific™ Nunc™ EasyFill™ Cell Factory system. Each 10-layer, single-use system holds the equivalent of 36 T175 flasks.
Supplier: Thermo Scientific™ N140410
Description
Save valuable space with Thermo Scientific™ Nunc™ EasyFill™ Cell Factory system. Each 10-layer, single-use system holds the equivalent of 36 T175 flasks. The linear multi-layer cell culture format is easy to use and promotes easy scalability, ideal for both research and commercial-scale applications.
Features:
- 'Out of the box' system
- Large, vented screw closure
- Versatile port design for pouring and aseptic filling
- Fill and empty five times faster than T175 flasks
- No accessories required with EasyFill Cell Factory systems
To Get Started:
Simply pour the media directly into the EasyFill Cell Factory system's large opening.
Warranty: 90 days
Specifications
| EasyFill Cell Factory | |
| 6 Pk. | |
| Less waste, renewable material | |
| Vented | |
| 10 |
| 6/Cs. | |
| Cell Factory System | |
| Sterile | |
| 6320 cm² |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.