Learn More
Thermo Scientific™ Nunc™ Disposable Loops and Needles, needle
Catalog No. 13079B
Change view
Click to view available options
Quantity:
Carton of 600
Catalog No. Quantity
13079B Carton of 600
1 options
Catalog No. 13079B Supplier Thermo Scientific™ Supplier No. 254399
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
  • Loops available in 10 and 1μL sizes
  • Meet semi-quantitative standard for handling samples
  • Calibration documents available for any batch upon request
Specifications
Disposable Single-use
Sterility Sterile
Material Polystyrene
Color Yellow
Product Type Inoculating Needle
Quantity Carton of 600