Learn More
Thermo Scientific™ Nunc™ Cryobank and Bank-It™ Replacement Caps
Ensure contamination-free cap replacement. Closures are supplied racked for manual or automated replacement.
Supplier:  Thermo Scientific™ 374019
 View more versions of this product
Catalog No. 374019
Only null left
Description
Description
Ensure contamination-free cap replacement with Thermo Scientific™ Nunc™ Cryobank and Bank-it™ Caps, which are supplied racked for manual or automated replacement. Available in natural, green, red or blue for simplified sample tracking.
Warranty: 90 days
  • Racked caps for automated or manual replacement of used caps
  • Racked in 96 microplate pattern for easy contamination free pick
Specifications
Chemical Identifiers
Polypropylene
2D Barcode
10 racks of 96/case
960
Nunc
Closure
Cryobank, Bank-It
96
0.5mL and 1.0mL Cryobank and Bank-it Tubes
Specifications
Polypropylene
Closure
2D Barcode
Red (Cap)
96
0.5mL and 1.0mL Cryobank and Bank-it Tubes
Red
Non-sterile
Cryobank, Bank-It
10 racks of 96/case
960
Nunc
Product Suggestions
Product Suggestions