Learn More
Thermo Scientific™ Nunc™ Cryobank and Bank-It™ Replacement Caps
GREENER_CHOICE
Catalog No. 374017
Change view
Click to view available options
Closure Color:
Blue
Green
Natural
Red
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color
374017 Natural
374018 Green
374019 Red
374021 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 374017 Supplier Thermo Scientific™ Supplier No. 374017
Ensure contamination-free cap replacement. Closures are supplied racked for manual or automated replacement.
Ensure contamination-free cap replacement with Thermo Scientific™ Nunc™ Cryobank and Bank-it™ Caps, which are supplied racked for manual or automated replacement. Available in natural, green, red or blue for simplified sample tracking.
Warranty: 90 days
  • Racked caps for automated or manual replacement of used caps
  • Racked in 96 microplate pattern for easy contamination free pick
Specifications
Coding 2D Barcode
Vial Type Cryobank, Bank-It
Packaging 10 racks of 96/case
For Use With (Equipment) 0.5mL and 1.0mL Cryobank and Bank-it Tubes
Quantity 960
Product Line Nunc
Sterility Irradiated
Type Closure
Closure Color Natural
Material Polypropylene