Learn More
Thermo Scientific™ Nunc™ TripleFlask Treated Cell Culture Flasks
Description
Perform cell culture expansion and experience 500 cm2 total growth area within the footprint of a standard flask using Thermo Scientific Nunc TripleFlask Treated Cell Culture Flasks. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems. These flasks are subjected to Nunclon Delta treatment, a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types.
Three layers of growth surface provide a total culture area of 500 cm2 in the same footprint as a standard Nunc 175 cm2 flask.
- Recommended working volume is 200 mL
- Promotes monolayer adherent cell culture
- Also available barcoded with large Code 128
- Nunclon Delta Certificate included in each case for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
Warranty: 90 days
Specifications
Specifications
| Barcode | Yes |
| Description | Nunclon Delta Treated TripleFlask, Filter Cap, Barcoded |
| Capacity (Metric) | 200 mL |
| Material | Polystyrene |
| Neck Style | Straight |
| No. per Case | 32/Cs. |
| No. per Pack | 1 Ea. |
| Product Type | Flask |
| Shape | Same external dimensions as the standard Nunc T175 with straight neck |
| Disposable | Single-use |
| Show More |
Frequently Asked Questions (FAQs)
The design of the Nunc TripleFlask does not allow for mechanical scraping. However, cells are easily harvested using protein digesting enzymes, such as trypsin, collagenase, and Pronase or by using non-enzymatic dissociation solutions.
The following is a document link regarding culturing techniques for Nunc TripleFlask:
https://assets.thermofisher.com/TFS-Assets/BID/Technical-Notes/culturing-technique-nunc-tripleflask-technical-note.pdf
This depends largely on the cell line and working conditions (considerations include headspace for gas exchange, supplements used, and optimal feeding schedule, among others) but we do offer guidelines for development purposes (0.3 to 0.5 mL per cm2) . Note that these volumes are approximate and may have to be modified for best performance. T25 Flask = 7 mL, T75 Flask = 25 mL, T175 Flask = 55 mL, T225 Flask = 70 mL, 300cm2 Flask = 100 mL, 500 cm2 TripleFlask = 200 mL, 35 mm round culture dish = 3 mL, 60 mm round culture dish = 5 mL, 100 mm round culture dish = 12 mL, 150 mm round culture dish = 35 mL, 4 (rectangular) well Multidish = 5 mL, 8 (rectangular) well Multidish = 3 mL, 4 (round) well Multidish = 1 mL, 6 well Multidish = 3 mL, 12 well Multidish = 2 mL, 24 well Multidish = 1 mL, 48 well Multidish = 0.5 mL.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.