Learn More
Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates.
Supplier:  Thermo Scientific™ 262146
 View more versions of this product
Catalog No. 12565213
Only null left
Description
Description
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates. The clear and U shaped wells are ideal for solution-based assays (e.g. agglutination assays) and common serial dilutions, allowing clean and easy pipetting. Available in a variety of options for surfaces, sterility, closures, and packaging sizes.
  • Clear polystyrene has excellent optical properties ideal for optical measurements
  • Thermo Scientific™ Nunclon™ Delta is a hydrophilic surface treatment whereas the non-treated polystyrene is more hydrophobic
  • Customized bar coding is available
Warranty: 90 days
Specifications
Chemical Identifiers
Standard
Nunc
Polystyrene
Non-treated
8 x 12
Microplate
96
Round Bottom (U-well)
Specifications
American National Standards (ANSI) (ANSI/SBS 1-2004)
Without Lid
135 Pk.
Clear
Polystyrene
Non-sterile
Round Bottom (U-well)
135
Standard
8 x 12
Nunc
Microplate
96
Non-treated
135
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.