Learn More
Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates

Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.
Supplier: Thermo Scientific™ 249944
Description
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
Features:
- Shared-wall design increases the well volume
- Wider wells and rounded bottom improve mixing
- Conical bottom well for optimal sample recovery
- Black and white plates available for fluorescent- and luminescent-based applications
- Low binding polypropylene for homogeneous assays and storage
Identifiants chimiques
| Standard | |
| 96-well Polypropylene Sample Processing & Storage Plates | |
| Low binding PP | |
| 96-well plate | |
| Polypropylene | |
| Non-treated | |
| Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
| Yes |
| Without Lid | |
| 8 x 12 | |
| Nunc | |
| ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
| 96 | |
| Yes | |
| For Research Use Only. Not for use in diagnostic procedures. | |
| ANSI/ SLAS Standard |
Spécifications
| Custom Barcoding Available | |
| Standard | |
| 96-well Polypropylene Sample Processing & Storage Plates | |
| Autoclavable | |
| Low binding PP | |
| Natural | |
| ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
| 450 μL | |
| 400 μL | |
| Non-treated | |
| Yes | |
| 120 | |
| 120/Cs. | |
| Yes | |
| ANSI/ SLAS Standard |
| Certified RNase and DNase Free. ANSI Compliant. | |
| Without Lid | |
| 8 x 12 | |
| 20 pk., 120 cs. | |
| Nunc | |
| 96-well plate | |
| Polypropylene | |
| 96 | |
| Non-sterile | |
| Round, Conical Bottom (V-well) | |
| Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
| 20 | |
| For Research Use Only. Not for use in diagnostic procedures. | |
| Certified Quality |