Learn More
Thermo Scientific™ Nunc™ 96 DeepWell™ Polystyrene Plates
Clone and replicate assays with these untreated polystyrene plates, suited for agglutination assays and sample collection.
$1010.43 - $1290.33
Specifications
| Format | Standard |
|---|---|
| Lid | Without Lid |
| Array | 8 x 12 |
| Packaging | 32 Cases, 4 Pk. |
| Product Line | Nunc |
| Catalog Number | Mfr. No. | Sterility | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sterility | Price | Quantity | |||||
|
12-565-552
|
Thermo Scientific™
278605 |
Non-sterile |
Case of 32 Each for $1,010.43
|
|
|||||
|
12-565-553
|
Thermo Scientific™
278606 |
Sterile |
Case of 32 Each for $1,290.33
|
|
|||||
Description
Store samples and perform agglutination assays with Thermo Scientific™ Nunc™ 96 DeepWell™ Polystyrene Plates, featuring transparent round bottom deep wells that hold up to 900μL per well.
- Alpha-numeric grid provides quick and easy sample identification
- Optimized for robotics and liquid handling systems
- Round-bottom wells for optimal sample recovery
- Non-treated hydrophobic surface
- Sealable with well caps (Part No. 278616) or sealing tape
- Customized bar coding is available
Specifications
| Standard | |
| 8 x 12 | |
| Nunc | |
| Microplate | |
| 96 | |
| Non-treated | |
| 32 | |
| 128 x 86 x 41 mm |
| Without Lid | |
| 32 Cases, 4 Pk. | |
| Clear | |
| Polystyrene | |
| 900 μL | |
| Round Bottom (U-well) | |
| 4 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.