Learn More
Thermo Scientific™ Nunc™ 384-Well Microplate, white, TC surface, Pack of 10
Catalog No. 12565343
Catalog No. 12565343 Supplier Thermo Scientific™ Supplier No. 164610
Only null left
Improve background, autofluorescence and crosstalk with Nunc 384-well polystyrene white microplates.
Improve background, autofluorescence and crosstalk with Thermo Scientific™ Nunc 384-Well Polystyrene Black and White Microplates.
  • Color: White
  • Sterility: Sterile
  • Lid: With lid
White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence.
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Lid With Lid
Culture Area 0.084 cm2/well
Array 16 x 24
Packaging 10 Pk., 30 Cases
Product Line Nunc
Color White
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 120 μL
No. of Wells 384
Volume (Metric) Working 10 to 100 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
No. per Case 30
No. per Pack 10
Show More Show Less