Learn More
Thermo Scientific™ 384 Well Black Plate, TC Surface, Pack of 10
Color:Black; Sterility:Sterile; Lid:With Lid
Supplier:  Thermo Scientific™ 164564
Catalog No. 12565344
Only null left
Description
Description
Improve background, autofluorescence and crosstalk with Thermo Scientific™ Nunc™ 384-Well Polystyrene Black and White Microplates.
  • Black microplates are recommended for fluorescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
Specifications
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
0.084 cm2/well
10 Pk., 30 Cases
Black
Polystyrene
384
Sterile
Flat Bottom (F-well)
10
With Lid
16 x 24
Nunc
Microplate
120 μL
10 to 100 μL
Cell Culture
30
Case of 30
Product Suggestions
Product Suggestions