missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate
Description
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample and reagent volume reduction providing cost savings and allowing more samples to be screened. Our plates are high quality for consistent and reliable results.
Nunc 384-well polystyrene plates have rounded square-well geometry, eliminating wicking.
- Clear Polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Customized bar coding is available
- Flexible working volumes; 10-100μL
Specifications
Specifications
| Barcode | Custom Barcoding Available |
| Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
| Format | Standard |
| Lid | With Lid |
| Culture Area | 0.084 cm2/well |
| Description | Non-Treated; w/ lid; Sterile; 10/30 |
| Array | 16 x 24 |
| Packaging | 10 Pk., 30 Cases |
| Product Line | Nunc |
| Color | Clear |
| Show More |