missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ 14mL Round-Bottom Tube
Description
Perform critical cellular applications with Thermo Scientific™ Nunc™ 14mL Round-Bottom Tubes, facilitated by the dual position vent/close cap for optimized gas exchange. For research use only
- Optimized gas exchange with dual position vent/close cap
- Encased in breathable packaging
- Certificates of conformity available for every lot
- For research use only
Specifications
Specifications
| Closure Material | LDPE |
| Certifications/Compliance | Mouse Embryo Assay (1-cell stage MEA) tested. Produced under ISO 13485 with full lot traceability. USP Class VI tested. Non-pyrogenic and sterile (SAL 10-6). |
| Closure Type | Dual Position Vent/Close Cap |
| Length (Metric) | 95 mm |
| Material | Polystyrene |
| Quantity | 10/Pk., 500/Cs. |
| Sterility | Sterile |
| Capacity (Metric) | 14 mL |
| Shape | Round |
| Closure Included | Yes |
| Show More |