Learn More
D-Mark Biosciences D-Mark Biosciences NUCLEOMAG 96 PCR 4X96 PREPS DFS Item
Supplier:  D-Mark Biosciences 7441004
NucleoMag 96 PCR (4x96) 4x 96 preps for the purification of PCR products - NucleoMag P-Beads, Elution Plates U-bottom, Self-adhering PE Foils, buffers no dangerous goods according transport regulations
Catalog No. NC0442472
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions