missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Nuclear Fast Red Soln., enhanced stability
Description
A histological counterstain. Nuclear Fast Red solution turns cell nuclei red and cytoplasm a pale pink.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Nuclear Fast Red Soln. |
| Packaging | Plastic bottle |
| Synonym | Kernechtrot Solution |
| Quantity | 500 mL |
| Physical Form | Liquid |
RUO – Research Use Only