Abcam Inc Nuclear fast red, nucleic acid stain, 5G
Catalog No. 502704677
MW 357.28 g/mol. Fast-acting biological stain that targets nucleic acids. Works more quickly than Hematoxylin (ab143166). Also suitable for the determination of calcium.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 357.28 g/mol. Fast-acting biological stain that targets nucleic acids. Works more quickly than Hematoxylin (ab143166). Also suitable for the determination of calcium.