Learn More
MP BIOMEDICALS Nuc-Off Nucleases and Nucleic Acid Removal Spray
Non-toxic spray designed to remove nucleases and DNA.
$318.92 - $965.97
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
|
MP112461450
|
MP BIOMEDICALS
112461450 |
450 mL |
Each for $318.92
|
|
|||||
|
MP112461451
|
MP BIOMEDICALS
112461451 |
1800 mL |
Each for $965.97
|
|
|||||
Description
Biomedicals Nuc-Off Nucleases and DNA Removal Spray is a ready-to-use solution for fast and cost-effective removal of RNase, DNase and DNA contamination.
This non-toxic spray-type solution can effectively remove nucleases and DNA from the surface of:
- laboratory consumables
- instruments
- other utensils
creating a clean environment for RNA or DNA-related experiments.
It is strongly recommended to regularly clean your:
- pipettors
- bench surfaces
- instrument exterior
Application Notes:
Nuc-Off Nucleases and DNA Removal Spray is intended for effective and safe elimination of Nucleases and DNA that may cause false positive or false negative experimental results.
Usage Statement:
Research Use Only (RUO)
Specifications
| Removal Agent |