missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals NS-398, 98%

$186.12

Specifications

Description Off-white
Content And Storage Ambient temperatures
Assay 98%
CAS 123653-11-2
Molecular Formula C13H18N2O5S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ67313LB0
View Documents Promotion Details
Thermo Scientific Chemicals
J67313LB0
5 mg
Each for $186.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Off-white
98%
C13H18N2O5S
N-[2-(Cyclohexyloxy)-4-nitrophenyl]-methanesulfonamide;
Ambient temperatures
123653-11-2
314.36g/mol
Solid
Videos
SDS
Documents

Documents

RUO – Research Use Only