UserName
Recombinant human IRS1 (600-1245) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.