missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NP-40 lysis buffer with glycerol (2X)

$152.38

Specifications

Color Colorless
Concentration 2X
Content And Storage Keep cold
Format Viscous Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ60143AK
View Documents Promotion Details
Thermo Scientific™
J60143AK
250 mL
Each for $152.38
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Colorless
2X
Keep cold
Viscous Liquid
Videos
SDS
Documents

Documents