missing translation for 'onlineSavingsMsg'
Learn More

NP-40 lysis buffer (2X)

Catalog No. p-7042515
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62805AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62805AK Supplier Thermo Scientific Chemicals Supplier No. J62805AK
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Quantity 250 mL
Format Liquid
Content And Storage Ambient temperatures
Color Colorless
Concentration 2X

RUO – Research Use Only