missing translation for 'onlineSavingsMsg'
Learn More

NP-40 lysis buffer (2X)

Catalog No. AAJ62805AK
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62805AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62805AK Supplier Thermo Scientific Chemicals Supplier No. J62805AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Color Colorless
Concentration 2X
Content And Storage Ambient temperatures
Format Liquid
Quantity 250 mL

RUO – Research Use Only