accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Facility Safety and Maintenance
Cleaning Supplies and Equipment
Cleaners and Disinfectants
Laboratory Surfactants and Wetting Agents
MilliporeSigma™ NP-40 Alternative, Calbiochem™,
Structure Search
Print
MilliporeSigma™ NP-40 Alternative, Calbiochem™,
Catalog No.
492016100ML
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
Packaging
492016100ML
100 mL
Glass Bottle
492016500ML
500 mL
Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No.
492016100ML
Supplier
MilliporeSigma™
Supplier No.
492016100ML
Sign In
or
Register
to check your price and availability.
$120.11
/
Each
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
9016-45-9
Molecular Formula
C19H32O3
Molecular Weight (g/mol)
308.46
MDL Number
MFCD00134080
InChI Key
BLXVTZPGEOGTGG-UHFFFAOYSA-N
Synonym
Nonylphenyl Polyethylene Glycol
PubChem CID
24773
IUPAC Name
2-[2-(4-nonylphenoxy)ethoxy]ethan-1-ol
SMILES
CCCCCCCCCC1=CC=C(OCCOCCO)C=C1
Specifications
Color
Undesignated
Quantity
100 mL
Packaging
Glass Bottle
Product Suggestions
Documents