missing translation for 'onlineSavingsMsg'
Learn More

Tragacanth, Powder, NF, Spectrum™ Chemical

Catalog No. 18607478
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607478 50 kg Poly Bottle
18607475 500 g Poly Bottle
18607476 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607478 Supplier Spectrum Chemical Mfg Cor Supplier No. TR10550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TR105, 9000-65-1

Spectrum™ Chemical Tragacanth, Powder, NF is used as an emulsifier, thickener, stabilizer, and texturant additive. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9000-65-1

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 50 kg
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Tragacanth, Powder